Product Details

SNP ID
rs147946254
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33290395 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTCCGGTGCTAGTCAAACAGGAG[C/T]TGGGGGAGGCTCGGGCCACAGTAGG
Phenotype
MIM: 605660 MIM: 602306 MIM: 601962 MIM: 611440
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR6834 PubMed Links

Gene Details

Gene
MIR6834
Gene Name
microRNA 6834
There are no transcripts associated with this gene.

Gene
MIR6873
Gene Name
microRNA 6873
There are no transcripts associated with this gene.

Gene
PFDN6
Gene Name
prefoldin subunit 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185181.2 324 Silent Mutation CTG,TTG L69L NP_001172110.1
NM_001265595.1 324 Silent Mutation CTG,TTG L69L NP_001252524.1
NM_001265596.1 324 Silent Mutation CTG,TTG L69L NP_001252525.1
NM_014260.3 324 Silent Mutation CTG,TTG L69L NP_055075.1
Gene
RGL2
Gene Name
ral guanine nucleotide dissociation stimulator like 2
There are no transcripts associated with this gene.

Gene
TAPBP
Gene Name
TAP binding protein (tapasin)
There are no transcripts associated with this gene.

Gene
WDR46
Gene Name
WD repeat domain 46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164267.1 324 Intron NP_001157739.1
NM_005452.5 324 Intron NP_005443.3
XM_017011484.1 324 Intron XP_016866973.1
XM_017011485.1 324 Intron XP_016866974.1
XM_017011486.1 324 Intron XP_016866975.1
XM_017011487.1 324 Intron XP_016866976.1

View Full Product Details