Product Details

SNP ID
rs148338986
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116692151 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATATTAGTTCCACTGTACCCATT[C/G]CAGAGGTATACTTTACCAAAATATG
Phenotype
MIM: 604545
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KPNA5 PubMed Links

Gene Details

Gene
KPNA5
Gene Name
karyopherin subunit alpha 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002269.2 347 Missense Mutation CCA,GCA P79A NP_002260.2
XM_011535805.1 347 Missense Mutation CCA,GCA P99A XP_011534107.1
XM_017010838.1 347 Missense Mutation CCA,GCA P99A XP_016866327.1
XM_017010839.1 347 Missense Mutation CCA,GCA P79A XP_016866328.1
XM_017010840.1 347 Missense Mutation CCA,GCA P76A XP_016866329.1
XM_017010841.1 347 Intron XP_016866330.1
XM_017010842.1 347 UTR 5 XP_016866331.1
XM_017010843.1 347 UTR 5 XP_016866332.1

View Full Product Details