Product Details

SNP ID
rs149577265
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:34293504 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCACTTCAGTGACAATGAGCACA[C/T]AGACATACGTCCTGTGCTTCCTCTC
Phenotype
MIM: 609228
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUDT3 PubMed Links

Gene Details

Gene
NUDT3
Gene Name
nudix hydrolase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006703.3 738 Missense Mutation TAT,TGT Y96C NP_006694.1
Gene
RPS10-NUDT3
Gene Name
RPS10-NUDT3 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202470.2 738 Missense Mutation TAT,TGT Y215C NP_001189399.1

View Full Product Details