Product Details

SNP ID
rs139750664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:87504377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCACTATTAAGTCTGCATTCTGGA[C/T]GGTGGACAGGCGGTGAGCAATCACA
Phenotype
MIM: 171050
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCB1 PubMed Links

Gene Details

Gene
ABCB1
Gene Name
ATP binding cassette subfamily B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000927.4 4202 Missense Mutation ATC,GTC I1237V NP_000918.2

View Full Product Details