Product Details

SNP ID
rs139989467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:97971901 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACGCAAAGCGCAAAGAGACTGAAC[C/T]TGGGGTGTGGAGTGATACAGACCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MGC72080 PubMed Links

Gene Details

Gene
MGC72080
Gene Name
MGC72080 pseudogene
There are no transcripts associated with this gene.

Gene
MIR5692A1
Gene Name
microRNA 5692a-1
There are no transcripts associated with this gene.

Gene
MIR5692C2
Gene Name
microRNA 5692c-2
There are no transcripts associated with this gene.

View Full Product Details