Product Details

SNP ID
rs144349602
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:101031697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACCAACCCTGAGATGACCTCGA[C/T]TGAGTCCAGTGTGACTTCAGACACT
Phenotype
MIM: 608424
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MUC17 PubMed Links

Gene Details

Gene
MUC17
Gene Name
mucin 17, cell surface associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040105.1 334 Missense Mutation ACT,ATT T94I NP_001035194.1

View Full Product Details