Product Details

SNP ID
rs145651438
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:135927348 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTATTTGAACGATAAGCCTATAGA[C/T]AACAGTCTGAAGCTGCAAGGGAGAC
Phenotype
MIM: 611050 MIM: 606484
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LUZP6 PubMed Links

Gene Details

Gene
LUZP6
Gene Name
leucine zipper protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128619.2 3291 Missense Mutation ATC,GTC I35V NP_001122091.2
Gene
MTPN
Gene Name
myotrophin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145808.3 3291 UTR 3 NP_665807.1

View Full Product Details