Product Details
- SNP ID
-
rs146466435
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:130269758 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACGTCCAGGCAGTCAAAGTGTTCTT[A/G]GAGTCCCAGGGAATTGCCTATTCCA
- Phenotype
-
MIM: 600688
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CPA2
PubMed Links
Gene Details
- Gene
- CPA2
- Gene Name
- carboxypeptidase A2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001869.2 |
262 |
Silent Mutation |
TTA,TTG |
L81L |
NP_001860.2 |
View Full Product Details