Product Details

SNP ID
rs148248825
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:106867963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCACGCACCGGAGCCCGGGCCA[A/G]ATCCACCTGGTGCAGCGGCACCCGC
Phenotype
MIM: 601232
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIK3CG PubMed Links

Gene Details

Gene
PIK3CG
Gene Name
phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282426.1 545 Silent Mutation CAA,CAG Q134Q NP_001269355.1
NM_001282427.1 545 Silent Mutation CAA,CAG Q134Q NP_001269356.1
NM_002649.3 545 Silent Mutation CAA,CAG Q134Q NP_002640.2
XM_005250443.3 545 Silent Mutation CAA,CAG Q134Q XP_005250500.1
XM_011516316.1 545 Silent Mutation CAA,CAG Q134Q XP_011514618.1
XM_011516317.1 545 Silent Mutation CAA,CAG Q134Q XP_011514619.1
XM_017012328.1 545 Silent Mutation CAA,CAG Q134Q XP_016867817.1

View Full Product Details