Product Details

SNP ID
rs149297064
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:156950241 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGACCCTCCGCAGCCGCCACCGC[A/C]GCTGCCCGGAAACGGGCGCTTTTAG
Phenotype
MIM: 611269
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NOM1 PubMed Links

Gene Details

Gene
NOM1
Gene Name
nucleolar protein with MIF4G domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138400.1 519 Silent Mutation GCA,GCC A168A NP_612409.1

View Full Product Details