Product Details

SNP ID
rs150499567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:23246867 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGTGAGAATTCAGCATGGAATGT[C/T]TCTACTATTTCCTGGGATTTCTGCT
Phenotype
MIM: 604368
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPNMB PubMed Links

Gene Details

Gene
GPNMB
Gene Name
glycoprotein nmb
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005340.1 662 Missense Mutation CTC,TTC L4F NP_001005340.1
NM_002510.2 662 Missense Mutation CTC,TTC L4F NP_002501.1
XM_005249578.2 662 Missense Mutation CTC,TTC L4F XP_005249635.1
XM_017011676.1 662 Missense Mutation CTC,TTC L4F XP_016867165.1
XM_017011677.1 662 Missense Mutation CTC,TTC L4F XP_016867166.1
XM_017011678.1 662 Missense Mutation CTC,TTC L4F XP_016867167.1
Gene
LOC101927890
Gene Name
uncharacterized LOC101927890
There are no transcripts associated with this gene.

View Full Product Details