Product Details

SNP ID
rs140076606
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:81443484 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGAGGTTAATTGAGTATCTCAAG[C/T]TCAAACCAGAGAACAGAAAATTTGT
Phenotype
MIM: 170715
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PMP2 PubMed Links

Gene Details

Gene
PMP2
Gene Name
peripheral myelin protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002677.3 Intron NP_002668.1

View Full Product Details