Product Details

SNP ID
rs142387931
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:100924992 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCATTTTCAAATAGAAGACTTT[A/G]CTCTCTGCTTGTGAAGCATTGGGGA
Phenotype
MIM: 601288
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
YWHAZ PubMed Links

Gene Details

Gene
YWHAZ
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein zeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135699.1 877 Intron NP_001129171.1
NM_001135700.1 877 Intron NP_001129172.1
NM_001135701.1 877 Intron NP_001129173.1
NM_001135702.1 877 Intron NP_001129174.1
NM_003406.3 877 Intron NP_003397.1
NM_145690.2 877 Intron NP_663723.1
XM_005251061.3 877 Silent Mutation AGC,AGT S114S XP_005251118.1
XM_005251063.3 877 Silent Mutation AGC,AGT S114S XP_005251120.1
XM_011517290.2 877 Intron XP_011515592.1
XM_017013810.1 877 Silent Mutation AGC,AGT S114S XP_016869299.1
XM_017013811.1 877 Silent Mutation AGC,AGT S114S XP_016869300.1
XM_017013812.1 877 Intron XP_016869301.1

View Full Product Details