Product Details

SNP ID
rs149339630
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:81444596 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGTGATCCTCTCTGCAGGGTTAC[C/G]ATGCTCTGCAAAGACAAGGTCATGC
Phenotype
MIM: 170715
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PMP2 PubMed Links

Gene Details

Gene
PMP2
Gene Name
peripheral myelin protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002677.3 350 Silent Mutation ATC,ATG I84M NP_002668.1

View Full Product Details