Product Details

SNP ID
rs149624410
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22690605 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCGTGGCGCTTGCGCTCGTCGCT[C/G]CGCGCAAACTTGCGCCCGCAGAACT
Phenotype
MIM: 602419
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
EGR3 PubMed Links

Gene Details

Gene
EGR3
Gene Name
early growth response 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199880.1 1341 Silent Mutation CGC,CGG R306R NP_001186809.1
NM_001199881.1 1341 Silent Mutation CGC,CGG R290R NP_001186810.1
NM_004430.2 1341 Silent Mutation CGC,CGG R344R NP_004421.2
XM_005273425.3 1341 Silent Mutation CGC,CGG R399R XP_005273482.1
XM_005273426.3 1341 Silent Mutation CGC,CGG R306R XP_005273483.1
XM_011544429.2 1341 Silent Mutation CGC,CGG R363R XP_011542731.1

View Full Product Details