Product Details

SNP ID
rs140320148
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:111327854 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGACCACCAGCATGATGGTGTTCA[G/T]GAGCAGTGAACTTGTGCAAGCTAAC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR2K2 PubMed Links

Gene Details

Gene
OR2K2
Gene Name
olfactory receptor family 2 subfamily K member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_205859.1 580 Missense Mutation ATG,CTG M194L NP_995581.1
XM_011518520.2 580 Missense Mutation ATG,CTG M194L XP_011516822.1

View Full Product Details