Product Details

SNP ID
rs142090161
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104569441 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTTTCTGGCAGTAAAGAAAAAG[C/G]TTTCCTTCTCTGGGTGTATGGTGCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR13C8 PubMed Links

Gene Details

Gene
OR13C8
Gene Name
olfactory receptor family 13 subfamily C member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004483.1 274 Missense Mutation CTT,GTT L92V NP_001004483.1

View Full Product Details