Product Details

SNP ID
rs142964065
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137065557 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTCTGGCCCAGGCGGCGCTGGC[A/G]CTCCTGCACTCGTTGCAGCTGCTGC
Phenotype
MIM: 612057
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SAPCD2 PubMed Links

Gene Details

Gene
SAPCD2
Gene Name
suppressor APC domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178448.3 Intron NP_848543.2
XM_011519180.2 Intron XP_011517482.1

View Full Product Details