Product Details
- SNP ID
-
rs143822854
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:5231681 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGCCTGAGAAGACATTCACCACCA[C/T]CCCAGGAGGGTGGCTGCTGGAATCT
- Phenotype
-
MIM: 600910
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
INSL4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12720] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- INSL4
- Gene Name
- insulin like 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002195.1 |
263 |
Missense Mutation |
ACC,ATC |
T53I |
NP_002186.1 |
View Full Product Details