Product Details

SNP ID
rs144109720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:111327836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGGCAAGAGGAGAATGCTGACCA[A/C]CAGCATGATGGTGTTCATGAGCAGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR2K2 PubMed Links

Gene Details

Gene
OR2K2
Gene Name
olfactory receptor family 2 subfamily K member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_205859.1 598 Missense Mutation ATG,GTG M200V NP_995581.1
XM_011518520.2 598 Missense Mutation ATG,GTG M200V XP_011516822.1

View Full Product Details