Product Details

SNP ID
rs144943839
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136364346 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTTGTCGCTGCCCGTGGTGTTCT[C/G]CCGGTCCTCCTCCCCCTGCCGCCAT
Phenotype
MIM: 607212 MIM: 609491
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CARD9 PubMed Links

Gene Details

Gene
CARD9
Gene Name
caspase recruitment domain family member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052813.4 1733 Missense Mutation CAG,GAG Q523E NP_434700.2
NM_052814.3 1733 Intron NP_434701.1
Gene
DNLZ
Gene Name
DNL-type zinc finger
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080849.2 1733 Intron NP_001074318.1
Gene
GPSM1
Gene Name
G-protein signaling modulator 1
There are no transcripts associated with this gene.

View Full Product Details