Product Details

SNP ID
rs145243810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34382769 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACAGATACCTTAGGCAGCCGAGG[A/T]AGGTAGGTGGTATACTTGTTGCAGC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C9orf24 PubMed Links

Gene Details

Gene
C9orf24
Gene Name
chromosome 9 open reading frame 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252195.1 599 Intron NP_001239124.1
NM_032596.3 599 Silent Mutation CTA,CTT L127L NP_115985.2
NM_147168.1 599 Intron NP_671697.1
NM_147169.2 599 Intron NP_671698.1
XM_005251616.4 599 Silent Mutation CTA,CTT L127L XP_005251673.1
XM_005251618.4 599 Intron XP_005251675.1
XM_011518055.2 599 Intron XP_011516357.1
Gene
KIAA1161
Gene Name
KIAA1161
There are no transcripts associated with this gene.

View Full Product Details