Product Details

SNP ID
rs149383501
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127507247 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCTTGGCGCGCCGTCGCTTTTC[C/T]GAGAGGGTGGCTGACTCCGGGGTGC
Phenotype
MIM: 614045 MIM: 610933
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FAM129B PubMed Links

Gene Details

Gene
FAM129B
Gene Name
family with sequence similarity 129 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035534.2 1870 Silent Mutation TCA,TCG S600S NP_001030611.1
NM_022833.3 1870 Silent Mutation TCA,TCG S613S NP_073744.2
XM_005252135.2 1870 Silent Mutation TCA,TCG S638S XP_005252192.2
XM_011518925.1 1870 Silent Mutation TCA,TCG S643S XP_011517227.1
Gene
LRSAM1
Gene Name
leucine rich repeat and sterile alpha motif containing 1
There are no transcripts associated with this gene.

View Full Product Details