Product Details

SNP ID
rs149938143
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:111327507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAGAGATGTTCGTGTGTTTGATG[C/T]AATGTTATTTTGCCCAGCAATTTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2K2 PubMed Links

Gene Details

Gene
OR2K2
Gene Name
olfactory receptor family 2 subfamily K member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_205859.1 927 Silent Mutation TTA,TTG L309L NP_995581.1
XM_011518520.2 927 Silent Mutation TTA,TTG L309L XP_011516822.1

View Full Product Details