Product Details

SNP ID
rs112944065
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:155284906 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCTCAAGGTAACCAAAAATGCCA[C/T]AGAACTCTCACTGAAATTTAAATTT
Phenotype
MIM: 300138
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLIC2 PubMed Links

Gene Details

Gene
CLIC2
Gene Name
chloride intracellular channel 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289.5 Intron NP_001280.3

View Full Product Details