Product Details

SNP ID
rs121918362
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:46501237 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATATTTTACATTCTTGACCACTTT[C/A]ATCCTTCAGTGTTTCCTTGCCTATG
Phenotype
MIM: 300573
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
ZNF674 PubMed Links

Gene Details

Gene
ZNF674
Gene Name
zinc finger protein 674
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039891.2 750 Nonsense Mutation GAA,TAA E118* NP_001034980.1
NM_001146291.1 750 Nonsense Mutation GAA,TAA E112* NP_001139763.1
NM_001190417.1 750 Nonsense Mutation GAA,TAA E113* NP_001177346.1
XM_011543941.2 750 Intron XP_011542243.1
XM_011543943.2 750 Intron XP_011542245.1
XM_011543944.2 750 Intron XP_011542246.1
XM_011543945.2 750 Intron XP_011542247.1
XM_017029728.1 750 Intron XP_016885217.1
XM_017029729.1 750 Intron XP_016885218.1
XM_017029730.1 750 Nonsense Mutation GAA,TAA E51* XP_016885219.1
XM_017029731.1 750 Intron XP_016885220.1
XM_017029732.1 750 Intron XP_016885221.1
XM_017029733.1 750 Intron XP_016885222.1

View Full Product Details