Product Details

SNP ID
rs138991758
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:152135081 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCAAAGACCAGCAGCATGCACTC[A/G]GAGGCTTCACTAAACAACAAAGGGA
Phenotype
MIM: 300343
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAGEA10 PubMed Links

Gene Details

Gene
MAGEA10
Gene Name
MAGE family member A10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011543.2 973 Silent Mutation TCC,TCT S180S NP_001011543.2
NM_001251828.1 973 Silent Mutation TCC,TCT S180S NP_001238757.1
NM_021048.4 973 Silent Mutation TCC,TCT S180S NP_066386.2
Gene
MAGEA10-MAGEA5
Gene Name
MAGEA10-MAGEA5 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204811.2 973 Intron NP_001191740.1

View Full Product Details