Product Details

SNP ID
rs139551661
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:73951898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGCCATGCATATATGCAAAAACT[A/C]TAGAGAGGAATCAGAATTTTATAAA
Phenotype
MIM: 300832
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
JPX PubMed Links

Gene Details

Gene
JPX
Gene Name
JPX transcript, XIST activator (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details