Product Details

SNP ID
rs145804200
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:63640902 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTTCTTGCTCTTCCCATGTACA[C/T]ATTGTAACTACTGAATTCCTACATT
Phenotype
MIM: 300429
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF9 PubMed Links

Gene Details

Gene
ARHGEF9
Gene Name
Cdc42 guanine nucleotide exchange factor 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173479.1 Intron NP_001166950.1
NM_001173480.1 Intron NP_001166951.1
NM_015185.2 Intron NP_056000.1
XM_005262249.2 Intron XP_005262306.1
XM_005262250.3 Intron XP_005262307.1
XM_005262251.1 Intron XP_005262308.1
XM_005262252.1 Intron XP_005262309.1
XM_011530890.1 Intron XP_011529192.1
XM_011530892.1 Intron XP_011529194.1
XM_017029363.1 Intron XP_016884852.1
XM_017029364.1 Intron XP_016884853.1
XM_017029365.1 Intron XP_016884854.1
XM_017029366.1 Intron XP_016884855.1
XM_017029367.1 Intron XP_016884856.1
XM_017029368.1 Intron XP_016884857.1
XM_017029369.1 Intron XP_016884858.1
XM_017029370.1 Intron XP_016884859.1
XM_017029371.1 Intron XP_016884860.1
XM_017029372.1 Intron XP_016884861.1
XM_017029373.1 Intron XP_016884862.1
XM_017029374.1 Intron XP_016884863.1
XM_017029375.1 Intron XP_016884864.1
XM_017029376.1 Intron XP_016884865.1
XM_017029377.1 Intron XP_016884866.1
XM_017029378.1 Intron XP_016884867.1
XM_017029379.1 Intron XP_016884868.1
XM_017029380.1 Intron XP_016884869.1

View Full Product Details