Product Details

SNP ID
rs147026513
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:38149827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCATCACCAGGGAGACATAGCGCT[C/G]TTTGTCCATGCCATGCTTATCCACT
Phenotype
MIM: 300187
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SRPX PubMed Links

Gene Details

Gene
SRPX
Gene Name
sushi repeat containing protein, X-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170750.1 1411 Missense Mutation CAG,GAG Q407E NP_001164221.1
NM_001170751.1 1411 Missense Mutation CAG,GAG Q368E NP_001164222.1
NM_001170752.1 1411 UTR 3 NP_001164223.1
NM_006307.4 1411 Missense Mutation CAG,GAG Q427E NP_006298.1
XM_017029893.1 1411 Intron XP_016885382.1

View Full Product Details