Product Details

SNP ID
rs148370051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:142203303 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCACACTCAGAATAATAATCAGGA[C/G]GCTGTTCTCGGGCATGCCCTCATCA
Phenotype
MIM: 300468
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MAGEC2 PubMed Links

Gene Details

Gene
MAGEC2
Gene Name
MAGE family member C2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016249.3 Intron NP_057333.1

View Full Product Details