Product Details

SNP ID
rs1059046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:79559458 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGAGGCTGCCATCAAGATGAGG[G/C]TGAGGGCCAGAGGGCACAGCCACAT
Phenotype
MIM: 178642
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
SFTPA2 PubMed Links

Gene Details

Gene
SFTPA2
Gene Name
surfactant protein A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098668.3 Intron NP_001092138.1
NM_001320813.1 Intron NP_001307742.1
NM_001320814.1 Intron NP_001307743.1
XM_005270128.3 Intron XP_005270185.1
XM_005270132.3 Intron XP_005270189.1
XM_011540124.1 Intron XP_011538426.1
XM_011540125.1 Intron XP_011538427.1
XM_017016608.1 Intron XP_016872097.1

View Full Product Details