Product Details
- SNP ID
-
rs1043441
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:38734959 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTGCCCGCCTTCTTGCCCCTTCCC[T/C]GCAGCCAGACCACCAGACACAGCCG
- Phenotype
-
MIM: 602245
MIM: 613569
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GTPBP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs9835] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GTPBP1
- Gene Name
- GTP binding protein 1
- Gene
- SUN2
- Gene Name
- Sad1 and UNC84 domain containing 2
View Full Product Details