Product Details

SNP ID
rs1061673
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:19282596 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGGAGTTTTATTCTTCATTTGG[T/G]GGTGACTCTTCTATTAGGTTTTTGT
Phenotype
MIM: 608477 MIM: 608478
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
AKR7A3 PubMed Links

Gene Details

Gene
AKR7A3
Gene Name
aldo-keto reductase family 7 member A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012067.2 1208 UTR 3 NP_036199.2
XM_011541046.2 1208 Intron XP_011539348.1
XM_017000714.1 1208 Intron XP_016856203.1
Gene
AKR7L
Gene Name
aldo-keto reductase family 7 like (gene/pseudogene)
There are no transcripts associated with this gene.

View Full Product Details