Product Details

SNP ID
rs1061404
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18593380 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCACACACACACACACACCCACT[G/A]GGGTGCACCCAAAGGTGGCCTCACG
Phenotype
MIM: 604237
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
C19orf60 PubMed Links

Gene Details

Gene
C19orf60
Gene Name
chromosome 19 open reading frame 60
There are no transcripts associated with this gene.

Gene
CRLF1
Gene Name
cytokine receptor like factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004750.4 1728 UTR 3 NP_004741.1
XM_011528422.2 1728 Intron XP_011526724.1
XM_011528423.2 1728 UTR 3 XP_011526725.1
XM_011528424.2 1728 UTR 3 XP_011526726.1

View Full Product Details