Product Details

SNP ID
rs11557150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62309226 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACTTAAACCATCTTCAGAAACAA[A/G]ATCTGTCACCCGTAGAGCTTAGAGT
Phenotype
MIM: 610650 MIM: 601033
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADRM1 PubMed Links

Gene Details

Gene
ADRM1
Gene Name
adhesion regulating molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281437.1 11145 Intron NP_001268366.1
NM_001281438.1 11145 Intron NP_001268367.1
NM_007002.3 11145 Intron NP_008933.2
NM_175573.2 11145 Intron NP_783163.1
XM_005260257.1 11145 Intron XP_005260314.1
XM_011528503.1 11145 Intron XP_011526805.1
XM_017027602.1 11145 Intron XP_016883091.1
XM_017027603.1 11145 Intron XP_016883092.1
XM_017027604.1 11145 Intron XP_016883093.1
XM_017027605.1 11145 Intron XP_016883094.1
XM_017027606.1 11145 Intron XP_016883095.1
XM_017027607.1 11145 Intron XP_016883096.1
XM_017027608.1 11145 Intron XP_016883097.1
XM_017027609.1 11145 Intron XP_016883098.1
XM_017027610.1 11145 Intron XP_016883099.1
Gene
LAMA5
Gene Name
laminin subunit alpha 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005560.4 11145 UTR 3 NP_005551.3
XM_006723796.3 11145 UTR 3 XP_006723859.1
XM_006723798.3 11145 Intron XP_006723861.1
XM_011528818.2 11145 UTR 3 XP_011527120.1
XM_011528819.2 11145 UTR 3 XP_011527121.1

View Full Product Details