Product Details

SNP ID
rs11648118
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:15014430 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGTCAAGGGTTAGTCACCAGAC[A/G]TTGCCAAGAAATACGAAAAGAGAAT
Phenotype
MIM: 614244
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR1972-1 PubMed Links

Gene Details

Gene
MIR1972-1
Gene Name
microRNA 1972-1
There are no transcripts associated with this gene.

Gene
PDXDC1
Gene Name
pyridoxal dependent decarboxylase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001285444.1 Intron NP_001272373.1
NM_001285445.1 Intron NP_001272374.1
NM_001285447.1 Intron NP_001272376.1
NM_001285448.1 Intron NP_001272377.1
NM_001285449.1 Intron NP_001272378.1
NM_001285450.1 Intron NP_001272379.1
NM_001324019.1 Intron NP_001310948.1
NM_001324020.1 Intron NP_001310949.1
NM_001324021.1 Intron NP_001310950.1
NM_015027.3 Intron NP_055842.2
XM_005255173.1 Intron XP_005255230.1
XM_005255176.2 Intron XP_005255233.1
XM_006720865.2 Intron XP_006720928.2
XM_017023059.1 Intron XP_016878548.1
XM_017023060.1 Intron XP_016878549.1
XM_017023061.1 Intron XP_016878550.1
XM_017023062.1 Intron XP_016878551.1
XM_017023063.1 Intron XP_016878552.1
XM_017023064.1 Intron XP_016878553.1
XM_017023065.1 Intron XP_016878554.1

View Full Product Details