Product Details

SNP ID
rs11746917
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:663134 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCGTCTGTGATCGGGCGATTCCG[G/A]TGACCGCTCGTCTGTGATCGGGTGA
Phenotype
MIM: 616475 MIM: 608773
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
CEP72 PubMed Links

Gene Details

Gene
CEP72
Gene Name
centrosomal protein 72
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018140.3 4946 Intron NP_060610.2
XM_005248322.3 4946 Intron XP_005248379.1
XM_011514063.1 4946 Intron XP_011512365.1
XM_011514064.2 4946 Intron XP_011512366.1
XM_017009626.1 4946 Intron XP_016865115.1
XM_017009627.1 4946 Intron XP_016865116.1
Gene
LOC101929895
Gene Name
uncharacterized LOC101929895
There are no transcripts associated with this gene.

Gene
TPPP
Gene Name
tubulin polymerization promoting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007030.2 4946 UTR 3 NP_008961.1
XM_005248237.3 4946 UTR 3 XP_005248294.2
XM_017008993.1 4946 UTR 3 XP_016864482.1
XM_017008994.1 4946 UTR 3 XP_016864483.1

View Full Product Details