Product Details

SNP ID
rs181246888
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:24271510 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCGCCCGAGGCGGACGGAACGTC[C/T]TTTTTTCTTGAGACGTGGTTAATTG
Phenotype
MIM: 614140
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POM121L9P PubMed Links

Gene Details

Gene
POM121L9P
Gene Name
POM121 transmembrane nucleoporin like 9, pseudogene
There are no transcripts associated with this gene.

Gene
SPECC1L
Gene Name
sperm antigen with calponin homology and coiled-coil domains 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145468.3 Intron NP_001138940.3
NM_001254732.2 Intron NP_001241661.2
NM_001254733.1 Intron NP_001241662.1
NM_015330.4 Intron NP_056145.4
Gene
SPECC1L-ADORA2A
Gene Name
SPECC1L-ADORA2A readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details