Product Details

SNP ID
rs180706669
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:193180171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTTCTTCAGAAATTAAAAAAATA[C/G]TTCTTGAATTTCTTTATGTGATGAG
Phenotype
MIM: 603018 MIM: 607393
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
B3GALT2 PubMed Links

Gene Details

Gene
B3GALT2
Gene Name
beta-1,3-galactosyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003783.3 2107 UTR 3 NP_003774.1
Gene
CDC73
Gene Name
cell division cycle 73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024529.4 2107 Intron NP_078805.3
XM_006711537.3 2107 Intron XP_006711600.1

View Full Product Details