Product Details

SNP ID
rs181383922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73114379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGACCTGACAATGTCCCTGTATT[A/G]TGGAATAGCTTGCAGGAGAAAATTT
Phenotype
MIM: 609233
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUDT13 PubMed Links

Gene Details

Gene
NUDT13
Gene Name
nudix hydrolase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001283014.1 216 Missense Mutation TAT,TGT Y5C NP_001269943.1
NM_001283015.1 216 Missense Mutation TAT,TGT Y5C NP_001269944.1
NM_001283016.1 216 UTR 5 NP_001269945.1
NM_001283017.1 216 UTR 5 NP_001269946.1
NM_001283019.1 216 UTR 5 NP_001269948.1
NM_015901.5 216 Missense Mutation TAT,TGT Y5C NP_056985.3
XM_017016033.1 216 Missense Mutation TAT,TGT Y5C XP_016871522.1

View Full Product Details