Product Details

SNP ID
rs181464159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:38363580 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTGATGGACTTCAGGAGGTGTT[T/C]GAGACCGATGTCTTTGGCCATTTTA
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
HSD17B7P2 PubMed Links

Gene Details

Gene
HSD17B7P2
Gene Name
hydroxysteroid 17-beta dehydrogenase 7 pseudogene 2
There are no transcripts associated with this gene.

View Full Product Details