Product Details

SNP ID
rs182066253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:274453 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCCAGGGGACCAATTAAGAAAC[T/A]GCTATTTTCAGAGCAACAAAAATAA
Phenotype
MIM: 611380
Polymorphism
T/A, Transversion substitution
Allele Nomenclature
Literature Links
DIP2C PubMed Links

Gene Details

Gene
DIP2C
Gene Name
disco interacting protein 2 homolog C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014974.2 5924 UTR 3 NP_055789.1
XM_005252426.3 5924 UTR 3 XP_005252483.1
XM_005252427.3 5924 UTR 3 XP_005252484.1
XM_005252428.4 5924 UTR 3 XP_005252485.1
XM_005252430.2 5924 UTR 3 XP_005252487.2
XM_011519428.2 5924 UTR 3 XP_011517730.1
XM_011519429.2 5924 UTR 3 XP_011517731.1
XM_011519430.2 5924 UTR 3 XP_011517732.1
XM_011519431.2 5924 UTR 3 XP_011517733.1
XM_011519432.1 5924 UTR 3 XP_011517734.1
XM_011519433.2 5924 UTR 3 XP_011517735.1
XM_017015977.1 5924 UTR 3 XP_016871466.1

View Full Product Details