Product Details

SNP ID
rs182639596
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:74706114 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGATTTGAGACAGATGGCATAAA[A/C]AAATATTGCTAGCTATACAATAAAT
Phenotype
MIM: 123691
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CRYZ PubMed Links

Gene Details

Gene
CRYZ
Gene Name
crystallin zeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130042.1 1161 UTR 3 NP_001123514.1
NM_001130043.1 1161 UTR 3 NP_001123515.1
NM_001134759.1 1161 UTR 3 NP_001128231.1
NM_001889.3 1161 UTR 3 NP_001880.2
XM_005270491.4 1161 UTR 3 XP_005270548.1
XM_011540747.1 1161 UTR 3 XP_011539049.1
XM_017000367.1 1161 UTR 3 XP_016855856.1
XM_017000368.1 1161 UTR 3 XP_016855857.1

View Full Product Details