Product Details

SNP ID
rs183864846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:50270029 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGAGTCTAAATGTCAATCAAATA[A/C]ATTTCATATAGATCATTGTCAAAAT
Phenotype
MIM: 614759 MIM: 156535
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CFAP53 PubMed Links

Gene Details

Gene
CFAP53
Gene Name
cilia and flagella associated protein 53
There are no transcripts associated with this gene.

Gene
MBD1
Gene Name
methyl-CpG binding domain protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204136.1 2072 Intron NP_001191065.1
NM_001204137.1 2072 Intron NP_001191066.1
NM_001204138.1 2072 Intron NP_001191067.1
NM_001204139.1 2072 Intron NP_001191068.1
NM_001204140.1 2072 Intron NP_001191069.1
NM_001204141.1 2072 Intron NP_001191070.1
NM_001204142.1 2072 Missense Mutation ATG,ATT M545I NP_001191071.1
NM_001204143.1 2072 Intron NP_001191072.1
NM_001204151.2 2072 Intron NP_001191080.1
NM_001323942.1 2072 Intron NP_001310871.1
NM_001323947.1 2072 Intron NP_001310876.1
NM_001323949.1 2072 Intron NP_001310878.1
NM_001323950.1 2072 Intron NP_001310879.1
NM_001323951.1 2072 Intron NP_001310880.1
NM_001323952.1 2072 Intron NP_001310881.1
NM_001323953.1 2072 Intron NP_001310882.1
NM_001323954.1 2072 Intron NP_001310883.1
NM_002384.2 2072 Intron NP_002375.1
NM_015844.2 2072 Intron NP_056669.2
NM_015845.3 2072 Intron NP_056670.2
NM_015846.3 2072 Intron NP_056671.2
NM_015847.3 2072 Intron NP_056723.2
XM_005258271.2 2072 UTR 3 XP_005258328.1
XM_006722456.2 2072 Intron XP_006722519.1
XM_011525991.1 2072 Intron XP_011524293.1
XM_011525993.2 2072 Intron XP_011524295.1
XM_011525994.2 2072 Intron XP_011524296.1
XM_011525998.2 2072 Intron XP_011524300.1
XM_011525999.1 2072 Intron XP_011524301.1
XM_011526001.1 2072 Intron XP_011524303.1
XM_011526002.1 2072 UTR 3 XP_011524304.1
XM_011526003.2 2072 Intron XP_011524305.1
XM_011526006.1 2072 Intron XP_011524308.1
XM_011526007.1 2072 UTR 3 XP_011524309.1
XM_017025751.1 2072 Intron XP_016881240.1
XM_017025752.1 2072 Intron XP_016881241.1
XM_017025753.1 2072 Intron XP_016881242.1
XM_017025754.1 2072 Intron XP_016881243.1
XM_017025755.1 2072 Intron XP_016881244.1
XM_017025756.1 2072 Intron XP_016881245.1
XM_017025757.1 2072 UTR 3 XP_016881246.1
XM_017025758.1 2072 Intron XP_016881247.1
XM_017025759.1 2072 Intron XP_016881248.1
XM_017025760.1 2072 Intron XP_016881249.1
XM_017025761.1 2072 Intron XP_016881250.1
XM_017025762.1 2072 Intron XP_016881251.1
XM_017025763.1 2072 UTR 3 XP_016881252.1
XM_017025764.1 2072 Intron XP_016881253.1
XM_017025765.1 2072 Intron XP_016881254.1
XM_017025766.1 2072 Intron XP_016881255.1
XM_017025767.1 2072 Intron XP_016881256.1
XM_017025768.1 2072 Intron XP_016881257.1
XM_017025769.1 2072 Intron XP_016881258.1
XM_017025770.1 2072 UTR 3 XP_016881259.1
XM_017025771.1 2072 Intron XP_016881260.1
XM_017025772.1 2072 Intron XP_016881261.1
XM_017025773.1 2072 UTR 3 XP_016881262.1
XM_017025774.1 2072 Intron XP_016881263.1
XM_017025775.1 2072 Intron XP_016881264.1
XM_017025776.1 2072 UTR 3 XP_016881265.1
XM_017025777.1 2072 Intron XP_016881266.1

View Full Product Details