Product Details

SNP ID
rs183603024
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:46183926 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTCTAGGAAACATTTATTGTTTA[T/C]ATGCAGATCCTAGAGACTTCTATTT
Phenotype
MIM: 608490
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
SLC38A1 PubMed Links

Gene Details

Gene
SLC38A1
Gene Name
solute carrier family 38 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077484.1 7277 UTR 3 NP_001070952.1
NM_001278387.1 7277 UTR 3 NP_001265316.1
NM_001278388.1 7277 UTR 3 NP_001265317.1
NM_001278389.1 7277 UTR 3 NP_001265318.1
NM_001278390.1 7277 Intron NP_001265319.1
NM_030674.3 7277 UTR 3 NP_109599.3
XM_011538784.2 7277 Intron XP_011537086.1
XM_011538786.2 7277 Intron XP_011537088.1
XM_011538787.2 7277 Intron XP_011537089.1
XM_017019990.1 7277 Intron XP_016875479.1

View Full Product Details