Product Details

SNP ID
rs183759731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:127846689 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAGTAGAATTGCAATTTTATTTT[T/C]CTCCAGAGAAGTTTTTCTTCAGTCC
Phenotype
MIM: 606017
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
POLR2D PubMed Links

Gene Details

Gene
POLR2D
Gene Name
RNA polymerase II subunit D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004805.3 1902 UTR 3 NP_004796.1

View Full Product Details