Product Details

SNP ID
rs184309267
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15742246 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACCACCTATACTGTCTTCACCC[C/G]CTTCCTCAGCCCAATCATTTTCAGT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR10H3 PubMed Links

Gene Details

Gene
OR10H3
Gene Name
olfactory receptor family 10 subfamily H member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013938.1 854 Missense Mutation CCC,CGC P285R NP_039226.1

View Full Product Details