Product Details

SNP ID
rs184896100
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29906860 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCAAGGCAAGAGGAAGGCAGGGG[G/T]AGGGTCAGAGGTCAGTCCTTGAAGA
Phenotype
MIM: 608947 MIM: 616667
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ASPHD1 PubMed Links

Gene Details

Gene
ASPHD1
Gene Name
aspartate beta-hydroxylase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181718.3 1289 Intron NP_859069.2
XM_017023107.1 1289 Intron XP_016878596.1
Gene
KCTD13
Gene Name
potassium channel tetramerization domain containing 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178863.4 1289 UTR 3 NP_849194.1
XM_011545783.2 1289 UTR 3 XP_011544085.1
XM_011545784.2 1289 UTR 3 XP_011544086.1
XM_017023104.1 1289 UTR 3 XP_016878593.1
XM_017023105.1 1289 Intron XP_016878594.1
XM_017023106.1 1289 Intron XP_016878595.1
Gene
SEZ6L2
Gene Name
seizure related 6 homolog like 2
There are no transcripts associated with this gene.

View Full Product Details