Product Details

SNP ID
rs185054720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43576171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCAACGGCCCTTCGCAGCGGGCG[C/A]GCTGTCAGACCTCAGTCTGGCGGCT
Phenotype
MIM: 603968 MIM: 607845
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
POLH PubMed Links

Gene Details

Gene
POLH
Gene Name
DNA polymerase eta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291969.1 25 UTR 5 NP_001278898.1
NM_001291970.1 25 UTR 5 NP_001278899.1
NM_006502.2 25 UTR 5 NP_006493.1
XM_005249186.3 25 UTR 5 XP_005249243.1
XM_011514698.2 25 UTR 5 XP_011513000.1
Gene
XPO5
Gene Name
exportin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020750.2 25 Intron NP_065801.1

View Full Product Details